Learn More
PLANT CELL TECHNOLOGY PLANT PRESERVATIVE MIXTURE PPM
Supplier:  PLANT CELL TECHNOLOGY 100
Encompass
Plant Preservative Mixture (PPM) 100 Ml. Plant Preservative Mixture (PPM), is a broad-spectrum biocide/fungicide for plant tissue culture. PPM™ is the ultimate solution to the never-ending struggle against microbial airborne, waterborne and endogenous contamination. PPM™ is a heat stable preservative/biocide which can be used to effectively prevent or reduces microbial contamination in plant tissue culture. At optimum doses, PPM™, which stands for Plant Preservative Mixture™, is an extremely effective reservative/Biocide, yet does not impair in vitro seed germination, callus proliferation and callus regeneration.Despite the most stringent use of sterile techniques, the contamination of plant cell and plant tissue cultures remain a persistent problem that can result in losses ranging from small number of cultures to the loss of whole batches.PPM™ prevents the germination of both bacteria and fungi spores. It is heat stable and therefore can be autoclaved with the media.
Catalog No. NC0823665
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.