Learn More
Biomatik Corporation Plant Ribulose-1, 5-Bisphosphate Carboxylase/Oxygenase (Rubisco) Elisa Kit
Supplier:  Biomatik Corporation EKC31458
Encompass_Preferred
Plant Ribulose-1, 5-Bisphosphate Carboxylase/Oxygenase (Rubisco) Elisa Kit for detecting Ribulose-1, 5-Bisphosphate Carboxylase/Oxygenase(Rubisco) in plant tissues. Species Reactivity: Plant. Detection Range: 3.12-800ug/ml. Sensitivity: 3.12ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6115