Learn More
Bioworld PLANT RNA/DNA PURIFICATION KIT
Catalog No. 50196924
Encompass_Preferred
For simultaneous isolation of total RNA and DNA from the same plant sample No phenol extraction; High quality DNA and RNA are purified simultaneously using the same spin column; No need to split the lysate; All sizes of RNA are recovere...
Catalog No. 50-196-924 Supplier Bioworld Supplier No. 107600691
For simultaneous isolation of total RNA and DNA from the same plant sample No phenol extraction; High quality DNA and RNA are purified simultaneously using the same spin column; No need to split the lysate; All sizes of RNA are recovere...