Learn More
Epoch Life Science Inc GenCatch™ Plant RNA Purification System (250 preps) Uniquely formulated buffers, shearing tubes, reinforced columns with collection tubes, along with protocols.
SDP
Catalog No. 501538099
Encompass_Preferred
Provides a simple, fast and cost-effective method to purify total RNA from plant (and fungal) samples. Comes with shearing tubes for simple and fast homogenization and filtration of tissue. Total RNA with a molecular size greater than 200 nucleotides (which excludes smaller RNA such as 5S and 5.8S RNA, and tRNA) can be isolated. Silica membrane technology. Up to 100 ug yield. 50 ul elution volume. 100 mg sample size. Additional buffer provided for handling plant tissue that contains sticky secondary metabolites. RNA isolated can be used for a variety of molecular manipulations such as Northern blotting, RT-PCR, PolyA+ RNA selection, cDNA synthesis, Primer extension, in vitro translation, etc. No phenol/chloroform extraction nor ethanol precipitation required. Used by labs around the world. 18 month shelf life.
Catalog No. 50-153-8099 Supplier Epoch Life Science Inc Supplier No. 1760250
Provides a simple, fast and cost-effective method to purify total RNA from plant (and fungal) samples. Comes with shearing tubes for simple and fast homogenization and filtration of tissue. Total RNA with a molecular size greater than 200 nucleotides (which excludes smaller RNA such as 5S and 5.8S RNA, and tRNA) can be isolated. Silica membrane technology. Up to 100 ug yield. 50 ul elution volume. 100 mg sample size. Additional buffer provided for handling plant tissue that contains sticky secondary metabolites. RNA isolated can be used for a variety of molecular manipulations such as Northern blotting, RT-PCR, PolyA+ RNA selection, cDNA synthesis, Primer extension, in vitro translation, etc. No phenol/chloroform extraction nor ethanol precipitation required. Used by labs around the world. 18 month shelf life.