Learn More
Zymo Research Corporation Quick-DNA Plant/Seed Miniprep Kit (50 Preps)
SDP
Catalog No. 501251608
Encompass_Preferred
The procedure is easy and can be completed in as little as 15 minutes: plant samples (150 mg each) are added directly to a ZR BashingBead™ Lysis Tube (2.0 mm) and rapidly and efficiently lysed by bead beating without the use of organic denaturants or proteinases. Polysaccharides and polyphenols/tannins are removed from the DNA using our ZymoSpin™ Technology, which includes the Zymo-Spin™ III-HRC filter, a PCR inhibitor removal column. The eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, etc.
Catalog No. 50-125-1608 Supplier Zymo Research Corporation Supplier No. D6020
The procedure is easy and can be completed in as little as 15 minutes: plant samples (150 mg each) are added directly to a ZR BashingBead™ Lysis Tube (2.0 mm) and rapidly and efficiently lysed by bead beating without the use of organic denaturants or proteinases. Polysaccharides and polyphenols/tannins are removed from the DNA using our ZymoSpin™ Technology, which includes the Zymo-Spin™ III-HRC filter, a PCR inhibitor removal column. The eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, etc.