Learn More
Percival Scientific Plant Tissue Culture Chamber, CU30L2, 9.6 cu. ft., low heat PetriClear LED lighting, condensation reducing airflow, 2 tiers, 10-44° C ±0.5°, IntellusUltra controller
SDP
Catalog No. NC3730425
Encompass_Preferred
The CU30L2 chamber is optimized for plant tissue culture in plates, dishes or flasks. PetriClear LEDs are dimmable from 10-100% output with up to 190 µmoles/m2/s irradiance @ 6" from the LEDs. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Specially designed airflow insulates shelf level experiments from heat generated by the underlying light fixture. Two tiers of epoxy coated steel wire shelving slide in and out easily on stainless steel rail assemblies. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs compressor life, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for additional options.
Catalog No. NC3730425 Supplier Percival Scientific Supplier No. CU30L2
Add to Cart
Add to Cart
Request a Quote
The CU30L2 chamber is optimized for plant tissue culture in plates, dishes or flasks. PetriClear LEDs are dimmable from 10-100% output with up to 190 µmoles/m2/s irradiance @ 6" from the LEDs. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Specially designed airflow insulates shelf level experiments from heat generated by the underlying light fixture. Two tiers of epoxy coated steel wire shelving slide in and out easily on stainless steel rail assemblies. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs compressor life, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for additional options.