Learn More
Sigma Aldrich Fine Chemicals Biosciences Plant Total Protein Extrac
Catalog No. 502904833
Encompass
The Plant Total Protein Extraction Kit is designed specifically for use in plant bioscience to extract a qualitative sample of all proteins from any type of plant species or tissue. The protocol does not require any ultracentrifugation or aqueous polymer two-phase partitioning (APTP). The kit includes two reagents a plant specific protease inhibitor cocktail and a new chaotropic reagent with increased solubilizing power to extract more hydrophobic proteins (C0356 Protein Extraction Reagent Type 4). Routine use of the reagent in sequential protein extraction of a plant tissue sample with fresh reagent removes nearly all protein from the tissue. The protease inhibitor cocktail is a mixture of protease inhibitors with broad specificity for the inhibition of serine cysteine metalloproteases aspartic and aminopeptidases. It contains pepstatin A 4-(2-aminoethyl)benzenesulfonyl fluoride (AEBSf) E-64 leupeptin 1 10-phenanthroline and bestatin. The cocktail has been demonstrated to be
Catalog No. 50-290-4833 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PE02301KT
Add to Cart
Add to Cart
The Plant Total Protein Extraction Kit is designed specifically for use in plant bioscience to extract a qualitative sample of all proteins from any type of plant species or tissue. The protocol does not require any ultracentrifugation or aqueous polymer two-phase partitioning (APTP). The kit includes two reagents a plant specific protease inhibitor cocktail and a new chaotropic reagent with increased solubilizing power to extract more hydrophobic proteins (C0356 Protein Extraction Reagent Type 4). Routine use of the reagent in sequential protein extraction of a plant tissue sample with fresh reagent removes nearly all protein from the tissue. The protease inhibitor cocktail is a mixture of protease inhibitors with broad specificity for the inhibition of serine cysteine metalloproteases aspartic and aminopeptidases. It contains pepstatin A 4-(2-aminoethyl)benzenesulfonyl fluoride (AEBSf) E-64 leupeptin 1 10-phenanthroline and bestatin. The cocktail has been demonstrated to be