Learn More
Medchemexpress LLC HY-N0031 5mg Medchemexpress, Plantamajoside CAS:104777-68-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN00315MG
Encompass_Preferred
Medchemexpress, HY-N0031 5mg Plantamajoside CAS:104777-68-6 Plantamajoside is a phenylpropanoid glycoside isolated from Plantago asiatica L.(Plantaginaceae). Plantamajoside has protective effects on LPS-induced acute lung injury (ALI) mice model. Plantamajoside has the potential for the treatment of pulmonary inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2771