Learn More
World Precision Instrument Plantar Test Analgesia Meter, heated
SDP
Catalog No. 502538770
Encompass
This unit, which is designed for testing narcotics and strong non-narcotic drugs, offers both Plantar (Hargreaves Method) and Tail Flick testing with a single unit. Either testing system is also available individually.
II-390G : In plantar mode, the visible light/heat source is directed at the paw or other desired body part, and in tail flick mode it is directed at the subjects’ tails. Test up to 12 mice or 6 rats simultaneously.
Catalog No. 50-253-8770 Supplier World Precision Instrument Supplier No. II390G
Add to Cart
Add to Cart
Request a Quote
This unit, which is designed for testing narcotics and strong non-narcotic drugs, offers both Plantar (Hargreaves Method) and Tail Flick testing with a single unit. Either testing system is also available individually.
II-390G : In plantar mode, the visible light/heat source is directed at the paw or other desired body part, and in tail flick mode it is directed at the subjects’ tails. Test up to 12 mice or 6 rats simultaneously.