Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Planting Supplies
SCI_ED

Catalog No. S98108 Shop All Luster Leaf Products, Inc. Products
Change view
Click to view available options
Product Type:
Water-resistant Label Marker Pen
Color:
Black
Quantity:
12/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color Quantity
S98108 Water-resistant Label Marker Pen Black 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S98108 Supplier Luster Leaf Products, Inc. Supplier No. 870

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Water-resistant Label Marker Pen
Color Black
For Use With (Application) Luster Leaf Planting Supplies
Includes Black ink
Weight (English) 0.5 lb.
Weight (Metric) 0.22 kg
Quantity 12/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.