Learn More
Diagnostic Biosystems PLAP Placental Alkaline Phosphatase (PLAP) (SP15)
Catalog No. 502407482
Encompass_Preferred
This antibody reacts with a membrane-bound isoenzyme (Regan and Nagao type) of placental alkaline phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. This antibody is highly specific to PLAP and shows no cross-reaction with other isoenzymes of alkaline phosphatases. It is useful in the identification of testicular germ cell tumors. Unlike germ cell tumors, PLAP-positive somatic cell tumors uniformly express epithelial membrane antigen (EMA).
Catalog No. 50-240-7482 Supplier Diagnostic Biosystems Supplier No. RMPD017
This antibody reacts with a membrane-bound isoenzyme (Regan and Nagao type) of placental alkaline phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. This antibody is highly specific to PLAP and shows no cross-reaction with other isoenzymes of alkaline phosphatases. It is useful in the identification of testicular germ cell tumors. Unlike germ cell tumors, PLAP-positive somatic cell tumors uniformly express epithelial membrane antigen (EMA).