Learn More
Biogenex Laboratories PLAP : PL8-F6 : 6mL
SDP
Supplier:  Biogenex Laboratories AM2285M
Encompass_Preferred
Human placental alkaline phosphatase (PLAP), 60-70kD oncofetalantigen, is a member of a family of membrane bound alkalinephosphatase enzymes and isoenzymes. PLAP and/or PLAP-likeisoenzymes have been found to be expressed by malignant tumors of germcell and non-germ cell origin. The antibody reacts with PLAP insyncytiotrophoblasts in placenta and also reacts with human germ celltumors.
Catalog No. 50-197-8823