Learn More
Thomas Scientific AbsorbMax Film, Black
SDP
Catalog No. 501458239
Encompass_Preferred
Ideal for Fluorescent Assays Light-blocking and absorbing properties protect light-sensitive samples Versatile can be applied to either top or bottom of plate Reduces stray light and well-to-well crosstalk Notched film edges aligned with plate bottom supports facilitates positioning Withstands temperatures from -40°C to 80°C AbsorbMax™ sealing films are 200 µm thick black vinyl adhesive sealing films for use with black solid and clear bottom 96-, 384-, and 1536-well microplates. The sealing films may be applied to either the top or bottom of the plate. A split backing aids in applying the sealing film properly. For top-reading fluorescence applications, apply the black sealing film to the plate bottom with clear-bottom wells. For bottom-reading fluorescence, use the black sealing film as a top seal. Film dimensions are 114.3 x 76.2 mm. Available non-sterile and sterile. Excel Scientific Sealing Film Selection Chart
Catalog No. 50-145-8239 Supplier Thomas Scientific Supplier No. 1230Y26
Ideal for Fluorescent Assays Light-blocking and absorbing properties protect light-sensitive samples Versatile can be applied to either top or bottom of plate Reduces stray light and well-to-well crosstalk Notched film edges aligned with plate bottom supports facilitates positioning Withstands temperatures from -40°C to 80°C AbsorbMax™ sealing films are 200 µm thick black vinyl adhesive sealing films for use with black solid and clear bottom 96-, 384-, and 1536-well microplates. The sealing films may be applied to either the top or bottom of the plate. A split backing aids in applying the sealing film properly. For top-reading fluorescence applications, apply the black sealing film to the plate bottom with clear-bottom wells. For bottom-reading fluorescence, use the black sealing film as a top seal. Film dimensions are 114.3 x 76.2 mm. Available non-sterile and sterile. Excel Scientific Sealing Film Selection Chart