Learn More
Sigma Aldrich Fine Chemicals Biosciences Plas Labs desiccator cabin1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences PL863CGA1EA
Encompass
Plas-Labs desiccators are designed to protect store or assist in moving critical materials and prevent contamination. They are dust and moisture free enclosures which safeguard valuable research specimens or sensitive electronic components.Under positive N2 gas flow they prevent moisture from penetrating the molecular structure of stored components. You may easily observe the status of your components without opening the door. The amber units are very effective for protection from harmful U.V. rays for light sensitive components.Features Unique Gasket Guard system prevents gaskets from taking a set thus destroying their effectivenessAll shelves have perforated slots (grooves) for optimum gas saturationUnits are all stackable with non-slip grip tight feetStandardization of construction means inter changeable shelvingAll chambers with two or more cubicles have a built-in cubicle dividerClosed cellular PVC gasketingUnits are constructed of optically c
Catalog No. 50-290-8590