Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plas Labs™ Model 870 Closed Loop Isolator Glove Box
GSA_VASDP

Catalog No. 11389124 Shop All Plas-Labs, Inc. Products
Change view
Click to view available options
Width (English):
82 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
11-389-124 82 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-389-124 Supplier Plas Labs™ Supplier No. 870CLC
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Includes

Two pairs white Hypalon™ ambidextrous gloves. Stainless-steel cart.

Completely sealed system with automatic negative pressure control

Effective containment of potentially hazardous toxins or organisms.

  • Maintains slight negative working pressure: -0.50 in. H2O
  • Internal atmosphere continually filtered by inlet/outlet HEPA filters rated at 99.9997% efficiency
  • Accommodates two pairs of glove ports; with white ambidextrous Hypalon™ gloves
  • Multiple stations can be coupled together
  • Easy-to-read control panel with digital controllers and LED status lights
  • Clear acrylic, nonshadowing construction with easy-to-clean corners
  • Bright fluorescent lights for well-lit working conditions
  • Manual house exhaust valve for rapid venting
  • Stainless-steel support cart with gas-tank support tracks

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Glove Box
Depth (English) 47 in.
Depth (English) Interior 35 in.
Depth (Metric) 119 cm
Depth (Metric) Interior 89 cm
Height (English) 53 in.
Height (English) Interior 27 in.
Height (Metric) 135 cm
Width (English) 82 in.
Width (English) Interior 56 in.
Width (Metric) 208 cm
Width (Metric) Interior 142 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.