Learn More
MSC Midwest Rake Floor Squeegee, 24 in, Flat Stiff, 1 ea
Catalog No. 502815739
Encompass
This squeegee is designed for applying floor coatings such as epoxy, urethanes, and other floor coatings to various surfaces. It features a durable polymer frame that is color-coded for easy identification and provides industry-leading precision and versatility.
  • A 24 inch blade offers superior coverage
  • Flat stiff design is perfect for a variety of coating tasks
  • Durable polymer frame
  • Color-coded for easy identification
  • Ideal for applying epoxy, urethanes, and other floor coatings
  • Threaded end for easy installation on standard threaded handles (sold separately)
Catalog No. 50-281-5739 Supplier MSC Supplier No. 28803419
This squeegee is designed for applying floor coatings such as epoxy, urethanes, and other floor coatings to various surfaces. It features a durable polymer frame that is color-coded for easy identification and provides industry-leading precision and versatility.
  • A 24 inch blade offers superior coverage
  • Flat stiff design is perfect for a variety of coating tasks
  • Durable polymer frame
  • Color-coded for easy identification
  • Ideal for applying epoxy, urethanes, and other floor coatings
  • Threaded end for easy installation on standard threaded handles (sold separately)