Learn More
Harrick Plasma Expanded Plasma Cleaner 115V
SDP
Catalog No. NC0301989
Encompass_Preferred
The PDC-001 Expanded Plasma Cleaner is our larger benchtop plasma cleaner, extensively used for nanoscale surface cleaning and surface chemistry modification. Maximum RF power of 30W; Adjustable RF power settings (LOW, MEDIUM, HIGH); 6in. diameter x 6.5in. length Pyrex chamber; Hinged door with viewing window; Active fan cooling; Integral switch for a vacuum pump; Metering valve to qualitatively control gas flow and chamber pressure; Three-way valve to quickly switch from introducing gas, venting and isolating the chamber; Size: 11in. H x 18in. W x 9in. D; Weight: 37 lb. REQUIRED: A separate vacuum pump (minimum pump speed 23 L/min, ultimate total pressure of 200 mTorr or less) is required to run the plasma process.
Catalog No. NC0301989 Supplier Harrick Plasma Supplier No. PDC001
Add to Cart
Add to Cart
Request a Quote
The PDC-001 Expanded Plasma Cleaner is our larger benchtop plasma cleaner, extensively used for nanoscale surface cleaning and surface chemistry modification. Maximum RF power of 30W; Adjustable RF power settings (LOW, MEDIUM, HIGH); 6in. diameter x 6.5in. length Pyrex chamber; Hinged door with viewing window; Active fan cooling; Integral switch for a vacuum pump; Metering valve to qualitatively control gas flow and chamber pressure; Three-way valve to quickly switch from introducing gas, venting and isolating the chamber; Size: 11in. H x 18in. W x 9in. D; Weight: 37 lb. REQUIRED: A separate vacuum pump (minimum pump speed 23 L/min, ultimate total pressure of 200 mTorr or less) is required to run the plasma process.