Learn More
MSC PLASMA CUT DP4 FACESHIELD
Catalog No. 5000673141
Encompass
This Sellstrom Face Shield & Headgear provides reliable eye and face protection with a green, shade 5 polycarbonate lens, specifically designed for plasma cutting applications. It features a flip-up shield and a universal hard hat adapter for versatility and ease of use, while the anti-fog coating ensures clear visibility during tasks.
  • Anti-fog coated window for clear visibility
  • Polycarbonate window material for durability
  • Flip-up shield for intermittent use
  • Universal hard hat adapter for secure attachment
  • Window thickness of 0.0400 in.
  • Window dimensions of 12 in. high by 9 in. wide
  • Meets CSA Z94.3 and ANSI Z87.1-2015 standards
Catalog No. 50-006-73141 Supplier MSC Supplier No. 31231582
This Sellstrom Face Shield & Headgear provides reliable eye and face protection with a green, shade 5 polycarbonate lens, specifically designed for plasma cutting applications. It features a flip-up shield and a universal hard hat adapter for versatility and ease of use, while the anti-fog coating ensures clear visibility during tasks.
  • Anti-fog coated window for clear visibility
  • Polycarbonate window material for durability
  • Flip-up shield for intermittent use
  • Universal hard hat adapter for secure attachment
  • Window thickness of 0.0400 in.
  • Window dimensions of 12 in. high by 9 in. wide
  • Meets CSA Z94.3 and ANSI Z87.1-2015 standards