Learn More
ACROBiosystems Human Plasma Kallikrein/KLKB1, His Tag (KL1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Ala 638 (Accession # P03952-1).
Supplier:  ACROBiosystems KL1H52H9100UG
Encompass_Preferred
Human Plasma Kallikrein, a serine protease, is synthesized in the liver and circulates in the plasma by binding to high molecular weight (HMW) kininogen or as a free zymogen. The enzyme cleaves Lys-Arg and Arg-Ser bonds. It activates, in a reciprocal reaction, factor XII after its binding to a negatively charged surface. It also releases bradykinin from HMW kininogen and may also play a role in the renin-angiotensin system by converting prorenin into renin.
Catalog No. 50-225-3292