Learn More
Bioss Plasma kallikrein light chain
SDP
Catalog No. 5000066442
Encompass_Preferred
Plasma kallikrein light chain Primary Antibody Unconjugated Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB IHC-P IHC-F IP IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-66442 Supplier Bioss Supplier No. BSM61563R
Plasma kallikrein light chain Primary Antibody Unconjugated Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB IHC-P IHC-F IP IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol