Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EUROTROL Plasma/Low Hb Control Level 1, 2 vials per box, 1 mL per vial, Control Value 70 mg/dL, Storage 2-8 °C, Open-Vial Stability 30 days

Catalog No. NC1412060
Encompass

The EUROTROL Plasma/Low Hb Control Level 1 is a validated hematology control material designed for in vitro diagnostic use, specifically to verify accuracy and precision in blood testing. This control is formulated as a physiological buffered solution that closely mimics human plasma, ensuring high reliability in point-of-care testing environments.

  • Product type: Hematology control
  • Control level: 1 (Low Control)
  • Intended use: In vitro diagnostic
  • System compatibility: HemoCue Plasma/Low Hb System
  • Material: Buffered solution with bovine hemoglobin
  • Control value: 70 mg/dL
  • Packaging: 2 vials per box
  • Volume: 1 mL per vial
  • Storage temperature (unopened): 2-8 °C
  • Open-vial stability: 30 days
  • CLIA status: CLIA Waived / Professional Use Only
  • Regulatory compliance: Meets Directive 98/79/EC

Catalog No. NC1412060 Supplier EUROTROL Supplier No. 139.001.002
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The EUROTROL Plasma/Low Hb Control Level 1 is a validated hematology control material designed for in vitro diagnostic use, specifically to verify accuracy and precision in blood testing. This control is formulated as a physiological buffered solution that closely mimics human plasma, ensuring high reliability in point-of-care testing environments.

  • Product type: Hematology control
  • Control level: 1 (Low Control)
  • Intended use: In vitro diagnostic
  • System compatibility: HemoCue Plasma/Low Hb System
  • Material: Buffered solution with bovine hemoglobin
  • Control value: 70 mg/dL
  • Packaging: 2 vials per box
  • Volume: 1 mL per vial
  • Storage temperature (unopened): 2-8 °C
  • Open-vial stability: 30 days
  • CLIA status: CLIA Waived / Professional Use Only
  • Regulatory compliance: Meets Directive 98/79/EC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.