Learn More
Abcam Plasma Membrane Protein E x traction Kit, 50 Tests
Catalog No. 502215262
Encompass_Preferred
Plasma membrane protein extraction kit provides optimized buffers and reagents for the effective extraction of plasma membrane proteins from mammalian tissues and cells. The kit offers the option to either extract total cellular membrane proteins or specifically purify plasma membrane proteins. - Cited in over 200 publications - High yield and purity – consistent yield with high purity (over 90%) - Suitable for western blotting, 2-D gels, enzyme analyses, and more
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5262 Supplier Abcam Supplier No. AB6540050TEST
Plasma membrane protein extraction kit provides optimized buffers and reagents for the effective extraction of plasma membrane proteins from mammalian tissues and cells. The kit offers the option to either extract total cellular membrane proteins or specifically purify plasma membrane proteins. - Cited in over 200 publications - High yield and purity – consistent yield with high purity (over 90%) - Suitable for western blotting, 2-D gels, enzyme analyses, and more
The product is subject to the following: Abcam Restricted Use Statement