Learn More
SCP Science Nitric Acid, PlasmaPURE, 67-70%, Poly, 2.5L
Catalog No. NC0456190
Encompass
PlasmaPUREacids are manufactured with trace metals equal to or less than 1 ppb. Used for environmental and industrial applications in ICP-AES and flame atomic absorption spectroscopy. Complete with a Certificate of Analysis with lot number, expiry date, & maximum specification for over 60 analytes - Complete documentation for audit purposes Refined for low trace metals analysis 2 expiry dates (up to 3 years unopened and 15 months opened) - Long shelf life for unopened bottles
Catalog No. NC0456190 Supplier SCP Science Supplier No. 250038175
PlasmaPUREacids are manufactured with trace metals equal to or less than 1 ppb. Used for environmental and industrial applications in ICP-AES and flame atomic absorption spectroscopy. Complete with a Certificate of Analysis with lot number, expiry date, & maximum specification for over 60 analytes - Complete documentation for audit purposes Refined for low trace metals analysis 2 expiry dates (up to 3 years unopened and 15 months opened) - Long shelf life for unopened bottles