Learn More
Norgen Biotek Corp Plasma/Serum Exosome Purify Kt
Catalog No. NC1885146
Encompass_Preferred
The Plasma/Serum Exosome Purification Kits provide a fast, reliable and convenient method to purify and enrich for pure intact exosomes from different plasma/serum sample volumes ranging from 50 µL to 10 mL. These kits also allow for the purification of intact extracellular vesicles (EVs) from different plasma/serum sample volumes, and these EVs are ready for any downstream application. The purification is based on Norgen’s proprietary resin.These kits provide a clear advantage over other available methods since they do not require any special instrumentation, ultracentrifugation, precipitation reagents or any protease treatments. More importantly, the purified exosomes will not be contaminated with any other RNA-binding proteins that may contaminate your exosomal RNA, which is essential if studying exosomal RNA gene expression.
Catalog No. NC1885146 Supplier Norgen Biotek Corp Supplier No. 57400
The Plasma/Serum Exosome Purification Kits provide a fast, reliable and convenient method to purify and enrich for pure intact exosomes from different plasma/serum sample volumes ranging from 50 µL to 10 mL. These kits also allow for the purification of intact extracellular vesicles (EVs) from different plasma/serum sample volumes, and these EVs are ready for any downstream application. The purification is based on Norgen’s proprietary resin.These kits provide a clear advantage over other available methods since they do not require any special instrumentation, ultracentrifugation, precipitation reagents or any protease treatments. More importantly, the purified exosomes will not be contaminated with any other RNA-binding proteins that may contaminate your exosomal RNA, which is essential if studying exosomal RNA gene expression.