Learn More
Norgen Biotek Corp Plasma/Serum RNA Purification Maxi Kit
Catalog No. NC1569974
Encompass_Preferred
This kit provides a fast, reliable and convenient method to purify and concentrate high quality, high purity and inhibitor-free cell-free circulating and exosomal RNA using a two column method. This kit can purify RNA from fresh or frozen serum or plasma samples prepared from blood collected on either EDTA or Citrate, from volumes ranging from 2 mL to 5 mL. Plasma samples prepared from blood collected on heparin should not be used.The first column will handle the large volume input of bodily fluids that is followed by a concentration on a mini column for a final elution of 50 µL to 100 µL. All components for the purification and concentration are provided in one convenient and fast kit for the easy processing of large input volumes of bodily fluids.The purified plasma/serum RNA is fully compatible with all downstream applications including NGS, and more.
Catalog No. NC1569974 Supplier Norgen Biotek Corp Supplier No. 56200
This kit provides a fast, reliable and convenient method to purify and concentrate high quality, high purity and inhibitor-free cell-free circulating and exosomal RNA using a two column method. This kit can purify RNA from fresh or frozen serum or plasma samples prepared from blood collected on either EDTA or Citrate, from volumes ranging from 2 mL to 5 mL. Plasma samples prepared from blood collected on heparin should not be used.The first column will handle the large volume input of bodily fluids that is followed by a concentration on a mini column for a final elution of 50 µL to 100 µL. All components for the purification and concentration are provided in one convenient and fast kit for the easy processing of large input volumes of bodily fluids.The purified plasma/serum RNA is fully compatible with all downstream applications including NGS, and more.