Learn More
Perkin Elmer US LLC 0-Slot Demountable Quartz Torch for ELAN 5000/6x00/9000/DRCs & NexION 300/350
Catalog No. 509045543
Encompass_Preferred
This demountable 0-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for organic solvents. Included in: PFA-Platinum HF resistant Sample Introduction Kit (N8142001), PFA-Sapphire HF-Resistant Sample Introduction Kit (N8142002), NexION 300/350Q/X/D Consumables Kit (N8140513), NexION 300/350S Consumables Kit (N8140514), ELAN DRC-e/9000 Spares Kit (N8120610), and ELAN DRC II Spares Kit (N8120611).
Catalog No. 50-904-5543 Supplier Perkin Elmer US LLC Supplier No. N8122006
This demountable 0-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flow and is recommended for organic solvents. Included in: PFA-Platinum HF resistant Sample Introduction Kit (N8142001), PFA-Sapphire HF-Resistant Sample Introduction Kit (N8142002), NexION 300/350Q/X/D Consumables Kit (N8140513), NexION 300/350S Consumables Kit (N8140514), ELAN DRC-e/9000 Spares Kit (N8120610), and ELAN DRC II Spares Kit (N8120611).