Learn More
Charter Medical Plasma Transfer Set, 24 in, Needle Adapter, Roller Clamp, Piercing Pin, 96 units per case
Catalog No. NC2328017
Encompass
The Plasma Transfer Set is a sterile, disposable device designed for the efficient transfer of blood and blood components. This versatile 24-inch set features a roller clamp for flow control, a piercing pin for container access, and a needle adapter for connection. It meets national safety standards for handling blood and supports sterile connection technology for closed-system operations. Ideal for quality control sampling, this set is perfect for preparing or processing blood components from flexible, non-vented containers.
  • Tubing length: 24 in
  • Flow control: Roller clamp
  • Connection type: Needle adapter
  • Container access: Piercing pin
  • Application: Blood and blood component transfer
  • Compatibility: Sterile connection devices
  • Intended use: Quality control sampling
  • Packaging: 96 units per case
Catalog No. NC2328017 Supplier Charter Medical Supplier No. 0322030
The Plasma Transfer Set is a sterile, disposable device designed for the efficient transfer of blood and blood components. This versatile 24-inch set features a roller clamp for flow control, a piercing pin for container access, and a needle adapter for connection. It meets national safety standards for handling blood and supports sterile connection technology for closed-system operations. Ideal for quality control sampling, this set is perfect for preparing or processing blood components from flexible, non-vented containers.
  • Tubing length: 24 in
  • Flow control: Roller clamp
  • Connection type: Needle adapter
  • Container access: Piercing pin
  • Application: Blood and blood component transfer
  • Compatibility: Sterile connection devices
  • Intended use: Quality control sampling
  • Packaging: 96 units per case