Learn More
Bioworld Plasmid DNA Isolation Solution I, 250 mL
Catalog No. 50196878
Encompass_Preferred
Plasmid DNA ISolation, Solution I Common buffer used for DNA isolation. Technical Information: Glucose (50mM)Tris-HCl, pH 8.0 (25mM)EDTA, pH 8.0 (10mM)
Catalog No. 50-196-878 Supplier Bioworld Supplier No. 107600281
Plasmid DNA ISolation, Solution I Common buffer used for DNA isolation. Technical Information: Glucose (50mM)Tris-HCl, pH 8.0 (25mM)EDTA, pH 8.0 (10mM)