Learn More
Bioworld Plasmid DNA Isolation Solution II, 250 mL
Catalog No. 50196880
Encompass_Preferred
Plasmid DNA Isolation, Solution II Common buffer used for DNA isolation Technical Information: NaOH (0.2N) SDS (1%)
Catalog No. 50-196-880 Supplier Bioworld Supplier No. 107600291
Plasmid DNA Isolation, Solution II Common buffer used for DNA isolation Technical Information: NaOH (0.2N) SDS (1%)