Learn More
Bioworld Plasmid DNA Isolation Solution I, 500 mL
Catalog No. 50196879
Encompass_Preferred
Plasmid DNA ISolation, Solution I Common buffer used for DNA isolation. Technical Information: Glucose (50mM)Tris-HCl, pH 8.0 (25mM)EDTA, pH 8.0 (10mM)
Catalog No. 50-196-879 Supplier Bioworld Supplier No. 107600282
Plasmid DNA ISolation, Solution I Common buffer used for DNA isolation. Technical Information: Glucose (50mM)Tris-HCl, pH 8.0 (25mM)EDTA, pH 8.0 (10mM)