Learn More
Bioworld Plasmid DNA Isolation Solution III, 500 mL
Catalog No. 50196883
Encompass_Preferred
Plasmid DNA Isolation, Solution III Common buffer used for DNA isolation Technical Information: 5M Potassium Acetate (60ml) Glacial Acetic Acid (11.5ml) H2O (28.5ml) Resulting solution is 3M with respect to potassium and 5M with respect to acetate.
Catalog No. 50-196-883 Supplier Bioworld Supplier No. 107600302
Plasmid DNA Isolation, Solution III Common buffer used for DNA isolation Technical Information: 5M Potassium Acetate (60ml) Glacial Acetic Acid (11.5ml) H2O (28.5ml) Resulting solution is 3M with respect to potassium and 5M with respect to acetate.