Learn More
Bioworld Plasmid DNA Isolation Solution II, 500 mL
Supplier:  Bioworld 107600292
Encompass_Preferred
Plasmid DNA Isolation, Solution II Common buffer used for DNA isolation Technical Information: NaOH (0.2N) SDS (1%)
Catalog No. 50-196-881