Learn More
Teknova Plasmid DNA Medium (PDM). 1000mL, Sterile. 2 pack.
SDP
Catalog No. 502387616
Encompass
0.79% Tryptone, 0.44% Yeast Extract, 1.00% Cerelose Anhydrous Dextrose (Glucose), 1.28% Disodium Phosphate Heptahydrate,0.30% Monopotassium Phosphate Anhydrous, 0.02% Magnesium Sulfate Anhydrous, 0.05% Ammonium Chloride
Catalog No. 50-238-7616 Supplier Teknova Supplier No. P08202PK
0.79% Tryptone, 0.44% Yeast Extract, 1.00% Cerelose Anhydrous Dextrose (Glucose), 1.28% Disodium Phosphate Heptahydrate,0.30% Monopotassium Phosphate Anhydrous, 0.02% Magnesium Sulfate Anhydrous, 0.05% Ammonium Chloride