Learn More
Epoch Life Science Inc GenCatch™ Plasmid DNA Miniprep Spin Kit (50 preps) Uniquely formulated buffers, reinforced columns with collection tubes, along with protocols.
SDP
Catalog No. 501538113
Encompass_Preferred
Provides a simple, fast and cost-effective method to purify plasmid DNA without phenol/chloroform extraction. It is based on binding of DNA to silica-based membranes in a chaotropic salt. High quality spin columns, collection tubes together with buffers and protocols. HIGHLIGHTS Fast: Up to 24 samples can be readily prepared in 30 minutes with minimum effort. Reliable: Used by labs around the world. References as far back as 2006 proves quality and customers' trust in our products and services. Flexible: Mini columns can be used in either centrifugation procedure or using a vacuum manifold. Convenient: No phenol extractions or ethanol precipitations required. Quality: DNA prepared by this kit is suitable for many downstream processes including PCR, ligation, restriction enzyme digestion, transformation, radioactive and fluorescent sequencingg, etc. 18 month shelf life when stored at RT. Typical Yield: 4-20ug of DNA from 2ml of overnight culture. Binding Capacity: 40ug.
Catalog No. 50-153-8113 Supplier Epoch Life Science Inc Supplier No. 2160050
Provides a simple, fast and cost-effective method to purify plasmid DNA without phenol/chloroform extraction. It is based on binding of DNA to silica-based membranes in a chaotropic salt. High quality spin columns, collection tubes together with buffers and protocols. HIGHLIGHTS Fast: Up to 24 samples can be readily prepared in 30 minutes with minimum effort. Reliable: Used by labs around the world. References as far back as 2006 proves quality and customers' trust in our products and services. Flexible: Mini columns can be used in either centrifugation procedure or using a vacuum manifold. Convenient: No phenol extractions or ethanol precipitations required. Quality: DNA prepared by this kit is suitable for many downstream processes including PCR, ligation, restriction enzyme digestion, transformation, radioactive and fluorescent sequencingg, etc. 18 month shelf life when stored at RT. Typical Yield: 4-20ug of DNA from 2ml of overnight culture. Binding Capacity: 40ug.