Learn More
MDI Membrane PLASMID DNA MINIPREP KIT 50RXN
Catalog No. 501132196
Encompass_Preferred
pDNA Miniprep Kit offers silica membrane based spin column technology with a validated, simple bind, wash and elute protocol that allows plasmid DNA isolation from 1-5 mL culture volume in less than 30 minutes.It gives pDNA yield upto 25 µg due to its high binding capacity.
Catalog No. 50-113-2196 Supplier MDI Membrane Supplier No. MIPKXXXXXXX0050
pDNA Miniprep Kit offers silica membrane based spin column technology with a validated, simple bind, wash and elute protocol that allows plasmid DNA isolation from 1-5 mL culture volume in less than 30 minutes.It gives pDNA yield upto 25 µg due to its high binding capacity.