Learn More
Alkali Scientific Plasmid DNA Spin Columns Polyp
SDP
Catalog No. 503028746
Encompass_Preferred
Alkali ScientificPlasmid DNA Spin Columns provide a fast reliable and efficient solution for extracting high-quality plasmid DNA Supplied with column tubes and collection tubes these spin columns streamline the purification process ensuring high yields and exceptional purity They are suitable for a wide range of applications including cloning sequencing and transfection and are compatible with various spin-type column kits The columns are designed to extract nucleic acids from diverse sample types such as tissues cells plants and blood delivering large fragment sizes with reproducible results Key features include DNase- and RNase-free construction ensuring sample integrity and human DNA-free assurance making them ideal for sensitive downstream applications Large Fragment Size and High Purity Ensures optimal performance in molecular biology workflows High Quality and Reproducibility Provides consistent yields across different sample types Broad Sample Compatibility Suitable for t
Catalog No. 50-302-8746 Supplier Alkali Scientific Supplier No. GE002
Alkali ScientificPlasmid DNA Spin Columns provide a fast reliable and efficient solution for extracting high-quality plasmid DNA Supplied with column tubes and collection tubes these spin columns streamline the purification process ensuring high yields and exceptional purity They are suitable for a wide range of applications including cloning sequencing and transfection and are compatible with various spin-type column kits The columns are designed to extract nucleic acids from diverse sample types such as tissues cells plants and blood delivering large fragment sizes with reproducible results Key features include DNase- and RNase-free construction ensuring sample integrity and human DNA-free assurance making them ideal for sensitive downstream applications Large Fragment Size and High Purity Ensures optimal performance in molecular biology workflows High Quality and Reproducibility Provides consistent yields across different sample types Broad Sample Compatibility Suitable for t