Learn More
Bioworld Plasmid MaxiPrep Kit, 4 Prep(s)
Catalog No. 50196946
Encompass_Preferred
For the rapid preparation of high yields of plasmid DNA Endotoxin-free plasmid DNA(<0.1 EU/µg plasmid) Isolate up to 1.5 mg of plasmid DNA from 150 mL bacterial cultures Fast and efficient spin column procedure FEATURES AND BENEFITSFast and easy processing - Rapid spin-column format allows for the processing of multiple samples in 1.5 hours. High binding capacity of columns - The binding capacity of the columns in the Plasmid DNA MaxiPrep Kit (Endotoxin Free) is 1.5 mg. Endotoxin-free plasmid DNA - DNA isolated using Norgen’s Plasmid DNA MaxiPrep Kit (Endotoxin Free) has endotoxin levelsof 0.1 EU / µg of plasmid DNA or less. High yield - Up to 1 mg of plasmid DNA can be purified from 100 mL of bacterial culture. Recovered plasmid DNA is suitable for downstream applications - Purified plasmid DNA is fully compatible with transfections, transformations, restriction enzyme digestions and sequencing.Ships overnight, ambient temperature.
Catalog No. 50-196-946 Supplier Bioworld Supplier No. 107600891
For the rapid preparation of high yields of plasmid DNA Endotoxin-free plasmid DNA(<0.1 EU/µg plasmid) Isolate up to 1.5 mg of plasmid DNA from 150 mL bacterial cultures Fast and efficient spin column procedure FEATURES AND BENEFITSFast and easy processing - Rapid spin-column format allows for the processing of multiple samples in 1.5 hours. High binding capacity of columns - The binding capacity of the columns in the Plasmid DNA MaxiPrep Kit (Endotoxin Free) is 1.5 mg. Endotoxin-free plasmid DNA - DNA isolated using Norgen’s Plasmid DNA MaxiPrep Kit (Endotoxin Free) has endotoxin levelsof 0.1 EU / µg of plasmid DNA or less. High yield - Up to 1 mg of plasmid DNA can be purified from 100 mL of bacterial culture. Recovered plasmid DNA is suitable for downstream applications - Purified plasmid DNA is fully compatible with transfections, transformations, restriction enzyme digestions and sequencing.Ships overnight, ambient temperature.