Learn More
Zymo Research Corporation ZymoPURE II™ Plasmid Maxiprep (10 preps)
SDP
Catalog No. NC0835051
Encompass_Preferred
The ZymoPURE II Plasmid Maxiprep kit provides the fastest and simplest method available to efficiently isolate up to 1.2 mg of transfection grade plasmid DNA from E. coli. Utilizing a modified alkaline lysis in conjunction with a novel binding system, the ZymoPURE II Plasmid Maxiprep kit can process up to 150 ml of culture in less than 20 minutes. This remarkable kit results in significantly more plasmid DNA while providing drastically reduced processing times. The plasmid DNA is bound onto a column with either a vacuum or a centrifuge instead of a lengthy gravity flow column. Additionally, there is no ethanol precipitation step required and the elution is performed using a microcentrifuge. The recovered plasmid DNA is highly concentrated (up to 3 g/l), EndoZero, vaccine-grade, and transfection-ready. The ZymoPURE II Plasmid Maxiprep kit contains colored buffers that permit error-free visualization and identification of complete bacterial cell lysis and neutralization.
Catalog No. NC0835051 Supplier Zymo Research Corporation Supplier No. D4202
The ZymoPURE II Plasmid Maxiprep kit provides the fastest and simplest method available to efficiently isolate up to 1.2 mg of transfection grade plasmid DNA from E. coli. Utilizing a modified alkaline lysis in conjunction with a novel binding system, the ZymoPURE II Plasmid Maxiprep kit can process up to 150 ml of culture in less than 20 minutes. This remarkable kit results in significantly more plasmid DNA while providing drastically reduced processing times. The plasmid DNA is bound onto a column with either a vacuum or a centrifuge instead of a lengthy gravity flow column. Additionally, there is no ethanol precipitation step required and the elution is performed using a microcentrifuge. The recovered plasmid DNA is highly concentrated (up to 3 g/l), EndoZero, vaccine-grade, and transfection-ready. The ZymoPURE II Plasmid Maxiprep kit contains colored buffers that permit error-free visualization and identification of complete bacterial cell lysis and neutralization.