Learn More
Thomson Instrument Company Plasmid+ Media, Sterile, 1L
SDP
Catalog No. NC0085590
Encompass_Preferred
Plasmid+ Enriched Media is an optimized microbial growth medium formulated to drive high-copy plasmid production with clean downstreams. When paired with Ultra Yield Flasks and AirOtop closures, customers have reported plasmid yield increases of up to 610%.Key features
•Optimized formulation for E. coli growth and high-copy plasmids
•Supports low endotoxin output to reduce downstream cleanup
•Consistent, reproducible results across culture runs
•Works on standard orbital shakers from bench to pilot volumes
•Made in the USA with reliable availability
Catalog No. NC0085590 Supplier Thomson Instrument Company Supplier No. 446300
Plasmid+ Enriched Media is an optimized microbial growth medium formulated to drive high-copy plasmid production with clean downstreams. When paired with Ultra Yield Flasks and AirOtop closures, customers have reported plasmid yield increases of up to 610%.Key features
•Optimized formulation for E. coli growth and high-copy plasmids
•Supports low endotoxin output to reduce downstream cleanup
•Consistent, reproducible results across culture runs
•Works on standard orbital shakers from bench to pilot volumes
•Made in the USA with reliable availability