Learn More
LUNA NANOTECH INC Plasmid Midiprep Kit, 50 Preps
Supplier:  LUNA NANOTECH INC NK11250
Encompass_Preferred
PuroSPIN™ Plasmid MIDIprep Kit, 50 Preps
PuroSPIN™ Plasmid MIDIprep Kit is designed to provide simple and easy-to-use method for extraction and purification of plasmid DNA from cultured bacterial cells. The purification technology is based on plasmid binding to silica spin-columns, which allows efficient removal of genomic DNA, protein, salt and other contaminants. Silica spin columns offer faster, simpler and safer alternative to traditional nucleic acid purification techniques such as phenol-chloroform extraction. PuroSPIN™ buffer formulations produce highly purified plasmid DNA product for downstream applications, such as transfection, enzymatic digestion, or PCR.
Features:
- PuroSPIN™ Plasmid MIDIprep Kit can be used to purify plasmid DNA from up to 100 mL of overnight bacterial culture.
- Extaction of up to 200 µg of plasmid DNA product.
- Purify plasmid DNA from large volumes of bacterial culture
- Rapi
Catalog No. 50-256-6943