Learn More
Omega Biotek Inc E.Z.N.A. Plasmid Mini Kit I, Q(capless) Spin, 200preps
Catalog No. NC9777560
Encompass
The E.Z.N.A.Plasmid Mini Kit I is designed to isolate up to 30 g of high quality plasmid DNA from 1 5 mL of bacterial cultures in 30 minutes or less. This kit uses a modified alkaline lysis method to lyse the cells and separate genomic DNA from plasmid DNA. Plasmid DNA purification is further simplified by using our HiBindmini column technology in three quick steps: bind, wash and elute. Purified plasmid DNA is ready for a wide variety of downstream applications including routine screening, restriction enzyme digestion, DNA sequencing, cloning, transformation and transfection. The E.Z.N.A.Plasmid DNA Mini Kit I is available with two different types of columns V-spin columns have an attached cap (D6943), while Q-spin columns are capless (D6942). The columns are otherwise identical in use and application. Either column can be used with both the vacuum or centrifugation protocol.
Catalog No. NC9777560 Supplier Omega Biotek Inc Supplier No. D694202
The E.Z.N.A.Plasmid Mini Kit I is designed to isolate up to 30 g of high quality plasmid DNA from 1 5 mL of bacterial cultures in 30 minutes or less. This kit uses a modified alkaline lysis method to lyse the cells and separate genomic DNA from plasmid DNA. Plasmid DNA purification is further simplified by using our HiBindmini column technology in three quick steps: bind, wash and elute. Purified plasmid DNA is ready for a wide variety of downstream applications including routine screening, restriction enzyme digestion, DNA sequencing, cloning, transformation and transfection. The E.Z.N.A.Plasmid DNA Mini Kit I is available with two different types of columns V-spin columns have an attached cap (D6943), while Q-spin columns are capless (D6942). The columns are otherwise identical in use and application. Either column can be used with both the vacuum or centrifugation protocol.