Learn More
Bioland Scientific LLC Plasmid Miniprep Column, 250 columns, DNA-binding matrix, 10-30 μg plasmid DNA, 1-5 mL culture, less than 20 min processing time
SDP
Catalog No. NC1418787
Encompass_Preferred
The Bioland Scientific Plasmid Miniprep Column (250 count) is designed for the fast and efficient purification of high-purity plasmid DNA from bacterial cultures. Utilizing a DNA-binding specific matrix, these spin columns facilitate the isolation of 10-30 μg of high-copy plasmid DNA from 1-5 mL of culture in under 20 minutes. The purified DNA is suitable for various downstream molecular biology applications.
  • Column type: Spin column
  • Quantity: 250 columns
  • Capacity: 10-30 μg plasmid DNA
  • Input culture volume: 1-5 mL bacterial culture
  • Purification principle: DNA-binding specific matrix
  • Processing time: Less than 20 min
  • Elution compatibility: Tris buffer or pure water
  • Downstream applications: Restriction digestions, PCR, automated sequencing, transfection, restriction mapping, library screening, gene therapy, genetic vaccinations
Catalog No. NC1418787 Supplier Bioland Scientific LLC Supplier No. COL0102
The Bioland Scientific Plasmid Miniprep Column (250 count) is designed for the fast and efficient purification of high-purity plasmid DNA from bacterial cultures. Utilizing a DNA-binding specific matrix, these spin columns facilitate the isolation of 10-30 μg of high-copy plasmid DNA from 1-5 mL of culture in under 20 minutes. The purified DNA is suitable for various downstream molecular biology applications.
  • Column type: Spin column
  • Quantity: 250 columns
  • Capacity: 10-30 μg plasmid DNA
  • Input culture volume: 1-5 mL bacterial culture
  • Purification principle: DNA-binding specific matrix
  • Processing time: Less than 20 min
  • Elution compatibility: Tris buffer or pure water
  • Downstream applications: Restriction digestions, PCR, automated sequencing, transfection, restriction mapping, library screening, gene therapy, genetic vaccinations