Learn More
LUNA NANOTECH INC Plasmid Miniprep Kit, 50 Preps
Supplier:  LUNA NANOTECH INC NK11150
Encompass_Preferred
PuroSPIN™ Plasmid Miniprep Kit, 50 Preps
PuroSPIN™ Plasmid Miniprep Kit is designed to provide simple and easy-to-use method for extraction and purification of plasmid DNA from cultured bacterial cells. The purification technology is based on plasmid binding to silica spin-columns, which allows efficient removal of genomic DNA, protein, salt and other contaminants. Silica spin columns offer faster, simpler and safer alternative to traditional nucleic acid purification techniques such as phenol-chloroform extraction. PuroSPIN™ buffer formulations produce highly purified plasmid DNA product for downstream applications, such as transfection, enzymatic digestion, or PCR.
Features:
- Easily purify plasmids from bacterial cultures using silica spin columns
- Simply extraction process with no need for phenol or chloroform
- Optimized buffer formulations to ensure efficient removal of RNA from plasmid samples
- Efficient removal
Catalog No. 50-256-6945