Learn More
LGC GENOMICS LLC (Lucigen) Plasmid-Safe ATP-Dependent DNase, 10,000 U @ 10 U/LDigest unwanted contaminating linear double-stranded DNA, without harming circular double-stranded DNA such as plasmids and fosmids.
Catalog No. NC1424105
Encompass
Plasmid-Safe™ ATP-Dependent DNase selectively removes contaminating bacterial chromosomal DNA from plasmid, cosmid, fosmid, and BAC clones or vector preparations.
This item is not returnable. View return policy
Catalog No. NC1424105 Supplier LGC GENOMICS LLC Supplier No. E3110K
Plasmid-Safe™ ATP-Dependent DNase selectively removes contaminating bacterial chromosomal DNA from plasmid, cosmid, fosmid, and BAC clones or vector preparations.