Learn More
Athens Research And Technology Inc Plasmin (Lyophilized), Native Human Plasma, ≥90% (SDS-PAGE), 1 mg
Catalog No. 50489888
Encompass_Preferred
Plasmin (Lyophilized) is a native protein purified from human plasma with ≥90% (SDS-PAGE), supplied lyophilized in a 1 mg pack. Molecular weight: 81 kDa. Plasmin, Human Plasma: 1 MG; Lyophilized from 20 mM NaP04, pH 7.4, + 10 mg/ml D-Mannitol and 10 mg/ml NaCl. MW 76,500-85,000; EC=1.71; Min. Activity: > 5 units/mg protein. Purity: > =95% by SDS-PAGE. Purified from human plasma. Common research applications include protease activity, enzyme characterization and biochemical studies.
Catalog No. 50-489-888 Supplier Athens Research And Technology Inc Supplier No. 1616161213L1MG
Plasmin (Lyophilized) is a native protein purified from human plasma with ≥90% (SDS-PAGE), supplied lyophilized in a 1 mg pack. Molecular weight: 81 kDa. Plasmin, Human Plasma: 1 MG; Lyophilized from 20 mM NaP04, pH 7.4, + 10 mg/ml D-Mannitol and 10 mg/ml NaCl. MW 76,500-85,000; EC=1.71; Min. Activity: > 5 units/mg protein. Purity: > =95% by SDS-PAGE. Purified from human plasma. Common research applications include protease activity, enzyme characterization and biochemical studies.