Learn More
Cusabio Technology LLC plasminogen activator tissue
Catalog No. 5000050786
Encompass_Preferred
The Rat Tissue-type Plasminogen Activator (t-PA) ELISA Kit is designed for the quantitative detection of PLAT in serum plasma and tissue homogenates using a sandwich assay format This kit detects PLAT with a sensitivity of 0 078 ng/mL and a dynamic range of 0 312 ng/mL to 20 ng/mL providing precise measurements for research applications The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours Absorbance is measured at 450 nm ensuring reliable and reproducible data Suitable for studies involving fibrinolysis thrombosis or vascular biology this kit offers a robust tool for analyzing t-PA levels in rat-derived biological samples
Catalog No. 50-000-50786 Supplier Cusabio Technology LLC Supplier No. CSBE07917R
The Rat Tissue-type Plasminogen Activator (t-PA) ELISA Kit is designed for the quantitative detection of PLAT in serum plasma and tissue homogenates using a sandwich assay format This kit detects PLAT with a sensitivity of 0 078 ng/mL and a dynamic range of 0 312 ng/mL to 20 ng/mL providing precise measurements for research applications The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours Absorbance is measured at 450 nm ensuring reliable and reproducible data Suitable for studies involving fibrinolysis thrombosis or vascular biology this kit offers a robust tool for analyzing t-PA levels in rat-derived biological samples