Learn More
Athens Research And Technology Inc Plasminogen, Native Human Plasma, ≥95% (SDS-PAGE), 1 mg
Catalog No. 50489891
Encompass_Preferred
Plasminogen is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied lyophilized in a 1 mg pack. Molecular weight: 90-92 kDa. Plasminogen, Human Plasma: 1 MG; Lyophilized from 20 mM Tris-HCl, pH 7.5, with 2 mM EDTA; MW 90,000-92,000; EC=1.71; Activity: > 120 units/mg protein; Purity: > =95% by SDS-PAGE. Common research applications include protease activity, enzyme characterization and biochemical studies.
Catalog No. 50-489-891 Supplier Athens Research And Technology Inc Supplier No. 16161612001MG
Plasminogen is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied lyophilized in a 1 mg pack. Molecular weight: 90-92 kDa. Plasminogen, Human Plasma: 1 MG; Lyophilized from 20 mM Tris-HCl, pH 7.5, with 2 mM EDTA; MW 90,000-92,000; EC=1.71; Activity: > 120 units/mg protein; Purity: > =95% by SDS-PAGE. Common research applications include protease activity, enzyme characterization and biochemical studies.