Learn More
Alpco Ltd Porcine Plasminogen ELISA
SDP
Supplier:  Alpco Ltd 41PLAPOE01
Encompass
The Porcine Plasminogen ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring Plasminogen in biological samples of pigs. Research Use Only. Not for Use in Diagnostic Procedures.
Plasminogen is a glycoprotein produced by the liver. It is the precursor for plasmin, which targets fibrin in the process of dissolution of fibrin blood clots. Plasminogen is present in plasma and most extravascular fluids. The important role of plasminogen in fibrinolytic system makes it an interesting marker for various diseases.
Catalog No. 50-751-3608