Learn More
Bioss Plasmodium falciparum
SDP
Catalog No. 5000065312
Encompass_Preferred
Plasmodium falciparum Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65312 Supplier Bioss Supplier No. BSM72370M
Plasmodium falciparum Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide