Learn More
ATCC Plasmodium falciparum; FCR-3/FMG
Catalog No. 502381440
Encompass
Plasmodium falciparum strain FCR-3/FMG was isolated from a clinical specimen from Fajara, Gambia. This strain is cultivated in human erythrocytes and can be used in infectious disease research and vector-borne disease research.
Catalog No. 50-238-1440 Supplier ATCC Supplier No. 30932
Plasmodium falciparum strain FCR-3/FMG was isolated from a clinical specimen from Fajara, Gambia. This strain is cultivated in human erythrocytes and can be used in infectious disease research and vector-borne disease research.